Learn More
sec-Butyl isothiocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 115.2 |
Chemical Name or Material | sec-Butyl isothiocyanate |