Learn More
SEC11A Mouse anti-Human, Unconjugated, Clone: 2B11G12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67379-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839975
  • 1236.20 SEK / 20µL
Estimated Shipment: 03-10-2022
to see stock.
Description
Description
The SEC11A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEC11A. This antibody recognizes human, mouse, rat antigen. The SEC11A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
SEC11A
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat, Human
ELISA, Immunohistochemistry, Western Blot
2B11G12
Unconjugated
20 μL
23478
IgG2b