missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SEC13 |
---|---|
Concentration | 0.22 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16834554
|
Proteintech
15397-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16824554
|
Proteintech
15397-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene belongs to the SEC13 family of WD-repeat proteins. It is a constituent of the endoplasmic reticulum and the nuclear pore complex. It has similarity to the yeast SEC13 protein, which is required for vesicle biogenesis from endoplasmic reticulum during the transport of proteins. Multiple alternatively spliced transcript variants have been found.Specifications
SEC13 | |
Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
P55735, Q5XFW8, Q9D1M0 | |
110379, 297522, 6396 | |
SEC13 Fusion Protein Ag7610 | |
Primary | |
-20°C | |
SEC13 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3S1231E, Protein SEC13 homolog, SEC13, SEC13 homolog (S. cerevisiae), SEC13 like protein 1, SEC13 related protein, SEC13L1, SEC13R | |
SEC13 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |