missing translation for 'onlineSavingsMsg'
Learn More

SEC13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15397-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824554

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene belongs to the SEC13 family of WD-repeat proteins. It is a constituent of the endoplasmic reticulum and the nuclear pore complex. It has similarity to the yeast SEC13 protein, which is required for vesicle biogenesis from endoplasmic reticulum during the transport of proteins. Multiple alternatively spliced transcript variants have been found.
Specifications

Specifications

SEC13
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P55735, Q5XFW8, Q9D1M0
D3S1231E, Protein SEC13 homolog, SEC13, SEC13 homolog (S. cerevisiae), SEC13 like protein 1, SEC13 related protein, SEC13L1, SEC13R
Rabbit
IgG
150 μL
-20°C
Primary
110379, 297522, 6396
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SEC13
Liquid
SEC13
SEC13 Fusion Protein Ag7610
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers