missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC24C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16073-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Interacts with HSP70 and can stimulate its ATPase activity. Stimulates the association between HSC70 and HIP.Specifications
SEC24C | |
Polyclonal | |
Unconjugated | |
SEC24C | |
KIAA0079, SEC24 related protein C, SEC24C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
218811, 685144, 9632 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P53992 | |
Sec24c | |
SEC24C Fusion Protein Ag9030 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |