missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC61G Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11147-2-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody can recognize endogenous SEC61G that migrates at a molecular mass of ∽8 kDa.
SEC61G is a subunit of the SEC61 complex, which also contains alpha (SEC61A) and beta (SEC61B) subunits. The SEC61 complex is the central component of the protein translocation apparatus on the endoplasmic reticulum membrane. The gene of SEC61G maps to chromosome 7p11.2, and encodes a 68-amino acid single-pass membrane protein.Specifications
SEC61G | |
Polyclonal | |
Unconjugated | |
SEC61G | |
Sec61 gamma subunit, SEC61G, SSS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20335, 23480, 689134 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P60059, P60060 | |
Sec61g | |
SEC61G Fusion Protein Ag1624 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |