Läs mer
Secalciferol, TRC
Produktkod. 30138303
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138303 5 mg
30135060 10 mg
30003847 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138303 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS2115005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55721-11-4
Molekylformel C27 H44 O3
Molekylvikt (g/mol) 416.64
Synonym 2,3-Heptanediol, 2-methyl-6-[(1R,3aS,4E,7aR)-octahydro-4-[(2Z)-2-[(5S)-5-hydroxy-2-methylenecyclohexylidene]ethylidene]-7a-methyl-1H-inden-1-yl]-, (3R,6R)-, 9,10-Secocholesta-5,7,10(19)-triene-3,24,25-triol, (3β,5Z,7E,24R)- (9CI), (3R,6R)-2-Methyl-6-[(1R,3aS,4E,7aR)-octahydro-4-[(2Z)-2-[(5S)-5-hydroxy-2-methylenecyclohexylidene]ethylidene]-7a-methyl-1H-inden-1-yl]-2,3-heptanediol, (24R)-24,25-Dihydroxyvitamin D3, (24R)-Hydroxycalcidiol, 24(R),25-Dihydroxycholecalciferol, 24,25-Dihydroxycholecalciferol, (24R)-24,25-Dihydroxycholecalciferol, 24R,25-Dihydroxyvitamin D3, K-DR, Osteo D, PRI 1201, Ro 21-5816, Secalciferol, Vitamin D3-(24R),25-dihydroxy
IUPAC-namn (3R,6R)-6-[(1R,3aS,4E,7aR)-4-[(2Z)-2-[(5S)-5-hydroxy-2-methylidenecyclohexylidene]ethylidene]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-1-yl]-2-methylheptane-2,3-diol
LEDER C[C@H](CC[C@@H](O)C(C)(C)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)CCC3=C)\CCC[C@]12C
Specifikationer
Kemiskt namn eller material 24R,25-Dihydroxyvitamin D3 (Secalciferol)
Kvantitet 5 mg
InChI formel InChI=1S/C27H44O3/c1-18-8-12-22(28)17-21(18)11-10-20-7-6-16-27(5)23(13-14-24(20)27)19(2)9-15-25(29)26(3,4)30/h10-11,19,22-25,28-30H,1,6-9,12-17H2,2-5H3/b20-10+,21-11-/t19-,22+,23-,24+,25-,27-/m1/s1
Formel vikt 416.329