Beskrivning
SECISBP2 Polyclonal antibody specifically detects SECISBP2 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifikationer
Specifikationer
| Antigen | SECISBP2 |
| Användningsområden | Western Blot, Immunoprecipitation |
| Klassificering | Polyclonal |
| Konjugera | APC |
| Utspädning | Western Blot, Immunoprecipitation |
| Formulering | PBS |
| Gene Alias | DKFZp686C09169, SBP2SECIS-binding protein 2, SECIS binding protein 2, selenocysteine insertion sequence binding protein 2, selenocysteine insertion sequence-binding protein 2 |
| Värd art | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 804 to 854 of human SECIS Binding Protein 2 using the numbering given in entry NP_076982.3 (GeneID 79048). |
| Reningsmetod | Immunogen affinity purified |
| Visa mer |