missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SECTM1 Recombinant Protein

Human SECTM1 full-length ORF recombinant protein with GST-tag at N-terminal

Brand:  Abnova™ H00006398-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180652

  • 4515.00 SEK / 10µg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a transmembrane and secreted protein with characteristics of a type 1a transmembrane protein. It is found in a perinuclear Golgi-like pattern and thought to be involved in hematopoietic and/or immune system processes.

  • Theoretical MW (kDa): 49.94
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: QNEGWDSPICTEGVVSVSWGENTVMSCNISNAFSHVNIKLRAHGQESAIFNEVAPGYFSRDGWQLQVQGGVAQLVIKGARDSHAGLYMWHLVGHQRNNRQVTLEVSGAEPQSAPDTGFWPVPAVVTAVFILLVALVMFAWYRCRCSQQRREKKFFLLEPQMKFAALRAGAQQGLSRASAELWTPDSEPTPRPLALVFKPSPLGALELLSPQPLFPYAADP

Best when used within three months from the date of receipt.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH17716
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
49.94
8
Glutathione Sepharose 4 Fast Flow
10 ug
QNEGWDSPICTEGVVSVSWGENTVMSCNISNAFSHVNIKLRAHGQESAIFNEVAPGYFSRDGWQLQVQGGVAQLVIKGARDSHAGLYMWHLVGHQRNNRQVTLEVSGAEPQSAPDTGFWPVPAVVTAVFILLVALVMFAWYRCRCSQQRREKKFFLLEPQMKFAALRAGAQQGLSRASAELWTPDSEPTPRPLALVFKPSPLGALELLSPQPLFPYAADP
K12
SECTM1
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
6398
SECTM1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SECTM1
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers