Learn More
Simport™ Scientific SecureSeal™ Thermal Adhesive Sealing Film for PCR Application
Simport™ Scientific SecureSeal™ Thermal Adhesive Sealing Film for PCR Application consists of 2.0 mil polyolefin film coated on one side with pressure sensitive acrylate adhesive, which does not interfere with cycle reactions.
POA
Description
Description
  • Low-auto fluorescence
  • Thermostable and functional from −70°C to +100°C
  • Certified RNAse, DNAse and DNA-free
  • DMSO resistant
  • Certifications
    RNase, Dnase and DNA-free