Learn More
Simport™ Scientific SecureSeal™ Thermal Adhesive Sealing Film for PCR Application
Simport™ Scientific SecureSeal™ Thermal Adhesive Sealing Film for PCR Application consists of 2.0 mil polyolefin film coated on one side with pressure sensitive acrylate adhesive, which does not interfere with cycle reactions.
Brand:  Simport™ Scientific T329-1
Additional Details : Weight : 1.25639kg
 View more versions of this product
Product Code. 15151895
  • 8770.00 SEK / Pack of 1000
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Low-auto fluorescence
  • Thermostable and functional from −70°C to +100°C
  • Certified RNAse, DNAse and DNA-free
  • DMSO resistant
  • Certifications
    RNase, Dnase and DNA-free
Specifications
Specifications
Yes
Transparent
Non-sterile
PCR
Polyolefin
Sealing Film