missing translation for 'onlineSavingsMsg'
Läs mer

Selenosulfuric Acid Dipotassium Salt, Technical Grade, TRC

Produktkod. 30424107
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424107 500 mg
30420772 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30424107 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS252500500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 15571-89-8
Molekylformel K2O3SSe
Molekylvikt (g/mol) 237.22
Synonym Dipotassium Selenosulfate, Potassium selenosulfate, Potassium selenosulfate (K2SeSO3)
LEDER O=S(O[K])([Se][K])=O

Specifikationer

Kemiskt namn eller material Selenosulfuric Acid Dipotassium Salt
Kvantitet 500 mg
InChI formel InChI=1S/2K.H2O3SSe/c;;1-4(2,3)5/h;;(H2,1,2,3,5)/q2*+1;/p-2
Formel vikt 237.8008
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.