Learn More
Selenourea, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Selenourea is used in the synthesis of selenium heterocycles, which exhibits anti-inflammatory and anti-tumor activity. It acts as an effective ligand and form complexes with transition metals and metalloids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Selenourea is used in the synthesis of selenium heterocycles, which exhibits anti-inflammatory and anti-tumor activity. It acts as an effective ligand and form complexes with transition metals and metalloids.
Solubility
Soluble in water, alcohol and ether.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Selenourea is used in the synthesis of selenium heterocycles, which exhibits anti-inflammatory and anti-tumor activity. It acts as an effective ligand and form complexes with transition metals and metalloids.
Solubility
Soluble in water, alcohol and ether.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∽210°C (decomposition) |
Assay Percent Range | 99+% metals basis |
Quantity | 1 g |
UN Number | UN3283 |
Sensitivity | Air, light, and moisture sensitive |
Solubility Information | Soluble in water,alcohol and ether. |
Formula Weight | 123.02 |
Percent Purity | 99% |
Physical Form | Powder |
Chemical Name or Material | Selenourea |