Läs mer
Selisistat, MedChemExpress
Produktkod. 16496528
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16496528 10 mg
16443476 25 mg
16439117 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16496528 Leverantör Medchem Express Leverantörsnummer HY15452
MedChemExpress Selisistat (EX-527) is a potent and selective SirT1 (Sir2 in Drosophila melanogaster) inhibitor with an IC50 of 123 nM for SirT1. Selisistat alleviates pathology in multiple animal and cell models of Huntington's disease.
Selisistat (EX-527) is a potent and selective SirT1 (Sir2 in Drosophila melanogaster) inhibitor with an IC50 of 123 nM for SirT1. Selisistat alleviates pathology in multiple animal and cell models of Huntington's disease.
Specifikationer
Kemiskt namn eller material Selisistat
Fysisk form Solid
CAS 49843-98-3
Färg Light Nude
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C13H13ClN2O
För användning med (applikation) Cancer-programmed cell death
Synonym EX-527
Löslighetsinformation DMSO : ≥ 100 mg/mL (402.07 mM)
LEDER O=C(C1C(NC2=C3C=C(Cl)C=C2)=C3CCC1)N
Molekylvikt (g/mol) 248.71
Formel vikt 248.71
Procent renhet 98.0%