Läs mer
SELP Mouse anti-Human, Clone: Psel.KO.2.5, Abnova™
Beskrivning
This gene encodes a 140 kDa protein that is stored in the alpha-granules of platelets and Weibel-Palade bodies of endothelial cells. This protein redistributes to the plasma membrane during platelet activation and degranulation and mediates the interaction of activated endothelial cells or platelets with leukocytes. The membrane protein is a calcium-dependent receptor that binds to sialylated forms of Lewis blood group carbohydrate antigens on neutrophils and monocytes. Alternative splice variants may occur but are not well documented. [provided by RefSeq]
Specifikationer
Specifikationer
| Antigen | SELP |
| Användningsområden | Flow Cytometry, Immunohistochemistry (PFA fixed), Immunoprecipitation |
| Klassificering | Monoclonal |
| Klona | Psel.KO.2.5 |
| Konjugera | Unconjugated |
| Beskrivning | Mouse monoclonal antibody raised against human SELP. |
| Utspädning | Flow Cytometry (1:1-1:50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user. |
| Formulering | In PBS, pH 7.4 (0.09% sodium azide). |
| Gene Alias | CD62/CD62P/FLJ45155/GMP140/GRMP/LECAM3/PADGEM/PSEL |
| Gensymboler | SELP |
| Visa mer |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.