missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELPLG Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23605-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a glycoprotein that functions as a high affinity counter-receptor for the cell adhesion molecules P-, E- and L- selectin expressed on myeloid cells and stimulated T lymphocytes. As such, this protein plays a critical role in leukocyte trafficking during inflammation by tethering of leukocytes to activated platelets or endothelia expressing selectins. This protein requires two post-translational modifications, tyrosine sulfation and the addition of the sialyl Lewis x tetrasaccharide (sLex) to its O-linked glycans, for its high-affinity binding activity. Aberrant expression of this gene and polymorphisms in this gene are associated with defects in the innate and adaptive immune response. Alternate splicing results in multiple transcript variants.Specifications
SELPLG | |
Polyclonal | |
Unconjugated | |
SELPLG | |
CD162, CLA, PSGL 1, PSGL1, selectin P ligand, SELPLG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6404 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14242 | |
SELPLG | |
SELPLG Fusion Protein Ag19394 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |