missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ SEMA3F Polyclonal Antibody
GREENER_CHOICE

Produktkod. 17876908
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17876908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17876908 Leverantör Invitrogen™ Leverantörsnummer PA5143760

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

The antibody detects a 95 kDa* protein corresponding to the apparent molecular mass of Sema-3F on SDS-PAGE immunoblots of adult mouse brain and liver.

The semaphorins are a family of proteins that are involved in signaling. All the family members have a secretion signal, a 500-amino acid sema domain, and 16 conserved cysteine residues (Kolodkin et al., 1993 [PubMed 8269517]). Sequence comparisons have grouped the secreted semaphorins into 3 general classes, all of which also have an immunoglobulin domain. The semaphorin III family, consisting of human semaphorin III (SEMA3A; MIM 603961), chicken collapsin, and mouse semaphorins A, D, and E, all have a basic domain at the C terminus. Chicken collapsin contributes to path finding by axons during development by inhibiting extension of growth cones (Luo et al., 1993 [PubMed 8402908]) through an interaction with a collapsin response mediator protein of relative molecular mass 62K (CRMP62) (Goshima et al., 1995 [PubMed 7637782]), a putative homolog of an axonal guidance associated UNC33 gene product (MIM 601168). SEMA3F is a secreted member of the semaphorin III family.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen SEMA3F
Användningsområden Western Blot
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gen SEMA3F
Genaccessionsnr. O88632, Q13275
Gene Alias LOW QUALITY PROTEIN: semaphorin-3F; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3 F; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, 3F; Sema III/F; Sema IV; Sema3f; Sema4; SemaIV; SEMA-IV; SEMAK; semaphorin 3F; semaphorin III/F; semaphorin IV; Semaphorin-3F
Gensymboler SEMA3F
Värd art Rabbit
Immunogen Semaphorin 3F synthetic peptide (coupled to carrier protein) corresponds to amino acids from the C-terminal region of human Sema-3F. This sequence is highly conserved in rat and mouse Sema-3F, and has low homology to other semaphorin family members.
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 20350, 315996, 6405
Målarter Human, Mouse, Rat
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.