missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEMA4F Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12474-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
SEMA4F has growth cone collapse activity against retinal ganglion-cell axons.Specifications
SEMA4F | |
Polyclonal | |
Unconjugated | |
SEMA4F | |
M SEMA, m Sema M, PRO2353, Sema M, Sema W, SEMA4F, SEMAM, Semaphorin 4F, Semaphorin M, Semaphorin W, SEMAW | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10505, 20355, 29745 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95754, Q9Z123, Q9Z143 | |
SEMA4F | |
SEMA4F Fusion Protein Ag3082 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |