missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semenogelin-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15916-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is the predomit protein in semen. The encoded secreted protein is involved in the formation of a gel matrix that encases ejaculated spermatozoa. The prostate-specific antigen (PSA) protease processes this protein into smaller peptides, with each possibly having a separate function. The proteolysis process breaks down the gel matrix and allows the spermatozoa to move more freely.Specifications
Semenogelin-1 | |
Polyclonal | |
Unconjugated | |
SEMG1 | |
Semenogelin 1, semenogelin I, Semenogelin-1, SEMG, SEMG1, SGI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6406 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04279 | |
SEMG1 | |
Semenogelin-1 Fusion Protein Ag8717 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |