Läs mer
Semicarbazide-13C,15N2 Hydrochloride, TRC
Produktkod. 30041085
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30041085 1 mg
17919739 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30041085 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS2582021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1173020-16-0
Molekylformel 13C H5 15N2 N O . Cl H
Molekylvikt (g/mol) 114.51
Synonym Hydrazine-1,2-15N2-carboxamide-13C, hydrochloride (1:1)
IUPAC-namn (^{15}N)azanylurea;hydrochloride
LEDER Cl.[15NH2][15NH][13C](=O)N
Specifikationer
Kemiskt namn eller material Semicarbazide hydrochloride-13C,15N2
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/CH5N3O.ClH/c2-1(5)4-3;/h3H2,(H3,2,4,5);1H/i1+1,3+1,4+1;
Formel vikt 114.0174
Procent renhet >95%