Läs mer
Tocris Bioscience™ Senexin A
GREENER_CHOICE
Produktkod. 18732168
Change view
10mg
Produktkod.
18732168 10mg
1 options
Produktkod. 18732168 Leverantör Tocris Bioscience™ Leverantörsnummer 4875/10
Cdk8 inhibitor
Senexin A is a cyclin-dependent kinase 8 (cdk8) inhibitor (IC50 = 280 nM). Also binds to cdk19 in an ATP-competitive manner. Inhibits p21-induced transcription. Reverses doxorubicin -induced tumor-promoting paracrine activities in vivo.
TRUSTED_SUSTAINABILITY
Specifikationer
CAS 1780390-76-2
Kvantitet 10 mg
Produkttyp Senexin A
Renhet >98%
Molekylvikt (g/mol) 310.78
Inhibitorer Senexin A
For Research Use Only