missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Series 15 00 Porosity Filter Plates

Brand:  Robu™ 15259

Additional Details : Weight : 0.03230kg

 View more versions of this product

Product Code. 16882766

  • 444.00 SEK / Pack of 10

Estimated Shipment: 17-04-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

1Pack
Filter Plate
3 mm
25 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers