Learn More
Robu™ Series 15 Filter Discs
Robu™ Series 15 Filter Discs feature a unique porous structure, which make them flexible and incredibly versatile for a variety of applications.
Brand: Robu™ 15201
Additional Details : Weight : 0.01000kg
Description
- Surfaces untreated
- Ground cylindrical
- The faces are press molded and may show thickness deviations
Specifications
Untreated surface | |
100 to 160 μm | |
3 ± 0.7 mm | |
20 mm |
Glass | |
Cylindrical | |
Filter Disc |