Learn More
Globe Scientific Serological Pipet Storage Rack
Product Code. 31921924 Shop All Globe Scientific Products
Change view
Click to view available options
Color:
Blue
Red
White
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31921924 White Each
31921938 Blue Each
31923616 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31921924 Supplier Globe Scientific Supplier No. 1685W
Spotlight Offers
Use promo code 28038
View all promotions
Beräknad leverans: 07-01-2027
Four angled compartments protect and organize glass or plastic serological pipets
  • Durable ABS construction for scratch resistance and easy cleaning
  • Strong magnets on both sides for secure mounting to most metal surfaces
  • Rack can stand on flat surfaces without mounting
  • Two included accessory attachments keep pipet fillers and bulbs within reach
  • Side windows allow for easy viewing of pipet size and inventory
Specifications
Autoclavable Not Autoclavable
Color White
Holds Serological Pipets
Material ABS with Mounting Magnets
Positions 4
Shape Rectangular
Stackable No
Sterility Non-sterile
Product Type Side-mounted Pipet Holder
Shelves 4 Tiers
Height (Metric) 395 mm
Array 1 x 4
Width (Metric) 85 mm