Läs mer
Globe Scientific Serological Pipet Storage Rack
Beskrivning
- Durable ABS construction for scratch resistance and easy cleaning
- Strong magnets on both sides for secure mounting to most metal surfaces
- Rack can stand on flat surfaces without mounting
- Two included accessory attachments keep pipet fillers and bulbs within reach
- Side windows allow for easy viewing of pipet size and inventory
Specifikationer
Specifikationer
| Autoklaveringsbar | Not Autoclavable |
| Färg | Blue |
| Håller | Serological Pipets |
| Material | ABS with Mounting Magnets |
| Positioner | 4 |
| Form | Rectangular |
| Staplingsbar | No |
| Sterilitet | Non-sterile |
| Produkttyp | Side-mounted Pipet Holder |
| Hyllor | 4 Tiers |
| Visa mer |