Läs mer
Serotonin Beta-D-Glucuronide, TRC
Produktkod. 17924255
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17924255 50 mg
17943135 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17924255 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS27499050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18186-43-1
Molekylformel C16H20N2O7
Molekylvikt (g/mol) 352.34
Synonym 3-(2-Aminoethyl)-1H-indol-5-yl β-D-Glucopyranosiduronic acid, 5-Hydroxytryptamine O-Glucuronide, 5-Hydroxytryptamine Glucuronide, Serotonin β-D-Glucuronide
IUPAC-namn (2S,3S,4S,5R,6S)-6-((3-(2-aminoethyl)-1H-indol-5-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid
LEDER O[C@H]1[C@H](O)[C@@H](O)[C@H](OC2=CC=C(NC=C3CCN)C3=C2)O[C@@H]1C(O)=O
Specifikationer
Kemiskt namn eller material Serotonin b-D-Glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C16H20N2O7/c17-4-3-7-6-18-10-2-1-8(5-9(7)10)24-16-13(21)11(19)12(20)14(25-16)15(22)23/h1-2,5-6,11-14,16,18-21H,3-4,17H2,(H,22,23)/t11-,12-,13+,14-,16+/m0/s1
Formel vikt 352.13
Procent renhet >95%