Läs mer
Serotonin Hydrogenoxalate, TRC
Produktkod. 30417048
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417048 100 mg
30411492 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30417048 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS275008100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3036-16-6
Molekylformel C10H12N2O .C2H2O4
Molekylvikt (g/mol) 266.25
Synonym Serotonin Oxalate, Serotonin Bioxalate, 5-Hydroxytryptamine Oxalate, 5-Hydroxytryptamine Bioxalate, 5-Hydroxytryptamine Binoxalate, 5-HT Oxalate, 3-(2-Aminoethyl)-indol-5-ol Oxalate, 3-(2-Aminoethyl)-indol-5-ol Oxalate (1:1) (Salt), 3-(2-Aminoethyl)-1H-indol-5-ol Ethanedioate (1:1) (Salt)
IUPAC-namn 3-(2-aminoethyl)-1H-indol-5-ol;oxalic acid
LEDER NCCc1c[nH]c2ccc(O)cc12.OC(=O)C(=O)O
Specifikationer
Kemiskt namn eller material Serotonin Hydrogenoxalate
Kvantitet 100 mg
InChI formel InChI=1S/C10H12N2O.C2H2O4/c11-4-3-7-6-12-10-2-1-8(13)5-9(7)10;3-1(4)2(5)6/h1-2,5-6,12-13H,3-4,11H2;(H,3,4)(H,5,6)
Formel vikt 266.0903