missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINA10 Mouse anti-Human, Porcine, Rat, Clone: 5B3C9, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SERPINA10 |
---|---|
Clone | 5B3C9 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887075
|
Proteintech
60113-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16877075
|
Proteintech
60113-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene belongs to the serpin family. It is predomitly expressed in the liver and secreted in plasma. It inhibits the activity of coagulation factors Xa and XIa in the presence of protein Z, calcium and phospholipid. Mutations in this gene are associated with venous thrombosis. Alternatively spliced transcript variants have been found for this gene.Specifications
SERPINA10 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PZI, Serpin A10, SERPINA10, ZPI | |
SERPINA10 | |
IgG2a | |
Protein A | |
Antibody |
5B3C9 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Porcine, Rat | |
Q62975, Q9UK55 | |
171154, 51156 | |
SERPINA10 Fusion Protein Ag2420 | |
Primary | |
-20°C | |
SERPINA10 |