missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SERPINA5 (Human) Recombinant Protein
Human SERPINA5 full-length ORF ( AAH08915, 17 a.a. - 406 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH08915 |
---|---|
Gene ID (Entrez) | 5104 |
Name | serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138771
|
Abnova™
H00005104-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16148771
|
Abnova™
H00005104-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: ASLHRHHPREMKKRVEDLHVGATVAPSSRRDFTFDLYRALASAAPSQNIFFSPVSISMSLAMLSLGAGSSTKMQILEGLGLNLQKSSEKELHRGFQQLLQELNQPRDGFQLSLGNALFTDLVVDLQDTFVSAMKTLYLADTFPTNFRDSAGAMKQINDYVAKQTKGKIVDLLKNLDSNAVVIMVNYIFFKAKWETSFNHKGTQEQDFYVTSETVVRVPMMSREDQYHYLLDRNLSCRVMGVPYQGNATALFILPSEGKMQQVENGLSEKTLRKWLKMFKKRQLELYLPKFSIEGSYQLEKVLPSLGISNVFTSHADLSGISNHSNIQVSEMVHKAVVEVDESGTRAAAATGTIFTFRSARLNSQRLVFNRPFLMFIVDNNILFLGRVNRP
Specifications
AAH08915 | |
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5 | |
125% SDS-PAGE Stained with Coomassie Blue | |
PAI3, PCI, PLANH3, PROCI | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5104 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SERPINA5 | |
GST |