missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SERPINA5 (Human) Recombinant Protein

Human SERPINA5 full-length ORF ( AAH08915, 17 a.a. - 406 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005104-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138771

  • 4515.00 SEK / 10µg
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: ASLHRHHPREMKKRVEDLHVGATVAPSSRRDFTFDLYRALASAAPSQNIFFSPVSISMSLAMLSLGAGSSTKMQILEGLGLNLQKSSEKELHRGFQQLLQELNQPRDGFQLSLGNALFTDLVVDLQDTFVSAMKTLYLADTFPTNFRDSAGAMKQINDYVAKQTKGKIVDLLKNLDSNAVVIMVNYIFFKAKWETSFNHKGTQEQDFYVTSETVVRVPMMSREDQYHYLLDRNLSCRVMGVPYQGNATALFILPSEGKMQQVENGLSEKTLRKWLKMFKKRQLELYLPKFSIEGSYQLEKVLPSLGISNVFTSHADLSGISNHSNIQVSEMVHKAVVEVDESGTRAAAATGTIFTFRSARLNSQRLVFNRPFLMFIVDNNILFLGRVNRP
Specifications

Specifications

AAH08915
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SERPINA5
GST
5104
Wheat germ expression system
10 μg
PAI3, PCI, PLANH3, PROCI
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers