missing translation for 'onlineSavingsMsg'
Learn More

SERPINB3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26558-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824985

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SPB3 may act as a protease inhibitor to modulate the host immune response against tumor cells.
Specifications

Specifications

SERPINB3
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P29508
HsT1196, Protein T4 A, SCC, SCCA, SCCA 1, SCCA PD, SCCA1, Serpin B3, SERPINB3, T4 A
Rabbit
IgG
20 μL
-20°C
Primary
6317
Western Blot
Unconjugated
SERPINB3
Liquid
SERPINB3
SERPINB3 Fusion Protein Ag24846
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers