missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SERPINB5 (Human) Recombinant Protein

Human SERPINB5 full-length ORF ( AAH20713, 1 a.a. - 231 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005268-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179061

  • 5030.41 SEK / 25µg

Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MDALQLANSAFAVDLFKQLCEKEPLGNVLFSPICLSTSLSLAQVGAKGDTANEIGQVLHFENVKDVPFGFQTVTSDVNKPSSFYSLKLIKRLYVDKSLNLSTEFISSTKRPYAKELETVDFKDKLEETKGQINNSIKDLTDGHFENILADNSVNDQTKILVVNAAYFVGKWMKKFPESETKECPFRVNRVCGAACSSKRSPIIDVKNDRDRVGHKSIPMRNLRARPAKCLS
Specifications

Specifications

AAH20713
serpin peptidase inhibitor, clade B (ovalbumin), member 5
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SERPINB5
GST
5268
Wheat germ expression system
25 μg
PI5, maspin
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates