missing translation for 'onlineSavingsMsg'
Learn More

SERPINB9 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14910-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16873814

  • 4721.67 SEK / 150µL
Estimated Shipment: 26-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PI9 belongs to the large superfamily of serine proteinase inhibitors (serpins), which bind to and inactivate serine proteinases. These interactions are involved in many cellular processes, including coagulation, fibrinolysis, complement fixation, matrix remodeling, and apoptosis (Sprecher et al., 1995 ).
Specifications

Specifications

SERPINB9
Polyclonal
Unconjugated
SERPINB9
CAP 3, CAP3, Cytoplasmic antiproteinase 3, Peptidase inhibitor 9, PI 9, PI9, Serpin B9, SERPINB9
Rabbit
Antigen Affinity Chromatography
RUO
5272
-20°C
Liquid
Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P50453
SERPINB9
SERPINB9 Fusion Protein Ag6707
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers