missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SERPINI2 (Human) Recombinant Protein
Human SERPINI2 full-length ORF ( NP_006208, 1 a.a. - 405 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005276-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MDTIFLWSLLLLFFGSQASRCSAQKNTEFAVDLYQEVSLSHKDNIIFSPLGITLVLEMVQLGAKGKAQQQIRQTLKQQETSAGEEFFVLKSFFSAISEKKQEFTFNLANALYLQEGFTVKEQYLHGNKEFFQSAIKLVDFQDAKACAEMISTWVERKTDGKIKDMFSGEEFGPLTRLVLVNAIYFKGDWKQKFRKEDTQLINFTKKNGSTVKIPMMKALLRTKYGYFSESSLNYQVLELSYKGDEFSLIIILPAEGMDIEEVEKLITAQQILKWLSEMQEEEVEISLPRFKVEQKVDFKDVLYSLNITEIFSGGCDLSGITDSSEVYVSQVTQKVFFEINEDGSEAATSTGIHIPVIMSLAQSQFIANHPFLFIMKHNPTESILFMGRVTNPDTQEIKGRDLDSL
Specifications
NP_006208 | |
serpin peptidase inhibitor, clade I (pancpin), member 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SERPINI2 | |
GST |
5276 | |
Wheat germ expression system | |
25 μg | |
MEPI, PANCPIN, PI14, TSA2004 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |