Läs mer
Sertaconazole Nitrate, TRC
Produktkod. 30138667
Change view
Kvantitet:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138667 10 mg
30135458 100 mg
17932945 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138667 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS27850010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 99592-39-9
Molekylformel C20 H15 Cl3 N2 O S . H N O3
Molekylvikt (g/mol) 500.78
Synonym Sertaconazole nitrate, (RS)-1-[2-[(7-Chloro-1-benzothiophen-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole nitrate, 1H-Imidazole, 1-[2-[(7-chlorobenzo[b]thien-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-, nitrate (1:1), 1H-Imidazole, 1-[2-[(7-chlorobenzo[b]thien-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-, mononitrate (9CI), Dermofix, Dermovit, Ertaczo, FI 7056, Sertaconazole mononitrate, Zalain
IUPAC-namn 1-[2-[(7-chloro-1-benzothiophen-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid
LEDER O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3csc4c(Cl)cccc34)c(Cl)c1
Specifikationer
Kemiskt namn eller material Sertaconazole Nitrate
Kvantitet 10 mg
InChI formel InChI=1S/C20H15Cl3N2OS.HNO3/c21-14-4-5-16(18(23)8-14)19(9-25-7-6-24-12-25)26-10-13-11-27-20-15(13)2-1-3-17(20)22;2-1(3)4/h1-8,11-12,19H,9-10H2;(H,2,3,4)
Formel vikt 498.9927