missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ SERTAD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535974
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (80%), Rat (83%), Rhesus Monkey (96%).
Acts at E2F-responsive promoters as coregulator to integrate signals provided by PHD- and/or bromodomain-containing transcription factors. May act as coactivator as well as corepressor of E2F1-TFDP1 and E2F4-TFDP1 complexes on E2F consensus binding sites, which would activate or inhibit E2F-target genes expression. Modulates fat storage by down-regulating the expression of key genes involved in adipocyte lipolysis, thermogenesis and oxidative metabolism. [UniProt].
Specifikationer
| SERTAD2 | |
| Polyclonal | |
| Unconjugated | |
| SERTAD2 | |
| AB041541; AU021878; Kiaa0127; MGC126688; MGC126690; mKIAA0127; MNCb-1504; PHD zinc finger- and bromodomain-interacting protein 2; Sei2; Sei-2; SERTA domain containing 2; SERTA domain-containing protein 2; SERTAD2; Transcriptional regulator interacting with the PHD-bromodomain 2; transcriptional regulator interacting with the PHS-bromodomain 2; TRIPBR2; TRIP-Br2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9792 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.74 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q14140 | |
| SERTAD2 | |
| Recombinant protein encompassing a sequence within the center region of human SERTAD2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering