Läs mer
Sertralinhydroklorid (1,0 mg/ml i metanol), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Sertraline Hydrochloride |
| CAS | 79559-97-0 |
| Molekylformel | C17 H17 Cl2 N . Cl H |
| Kvantitet | 5 x 1 mL |
| Synonym | Sertraline Hydrochloride, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride (1:1), (1S-cis)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthaleneamine hydrochloride, (1S,4S)-N-Methyl-4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrochloride, Altruline, Aremis, Atruline, Cp 51974-1, Dominum, Gladem, Jzoloft, Lesefer, Lustral, Selectra, Serlift, Sertraline hydrochloride, Seserine, Sirto, Sosser, Stimuloton, Zolof, Zoloft |
| InChI formel | InChI=1S/C17H17Cl2N.ClH/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11;/h2-6,8,10,12,17,20H,7,9H2,1H3;1H/t12-,17-;/m0./s1 |
| LEDER | Cl.CN[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13 |
| IUPAC-namn | (1S,4S)-4-(3,4-diklorofenyl)-N-metyl-1,2,3,4-tetrahydronafthalen-1-amin; hydroklorid |
| Molekylvikt (g/mol) | 342.69 |
| Formel vikt | 341.0505 |
| Visa mer |