missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24377-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
SETD9 is a protein-lysine N-methyltransferase. Monomethylates ′Lys-310′ of the RELA subunit of NF-kappa-B complex, leading to down-regulate NF-kappa-B transcription factor activity. Monomethylates ′Lys-8′ of H2AZ (H2AZK8me1). Required for the maintece of embryonic stem cell self-renewal.Specifications
SETD6 | |
Polyclonal | |
Unconjugated | |
SETD6 | |
SET domain containing 6, SETD6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
66083, 79918 | |
-20°C | |
Liquid |
Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TBK2, Q9CWY3 | |
Setd6 | |
SETD6 Fusion Protein Ag19485 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |