missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SETD6 |
---|---|
Concentration | 0.57 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882845
|
Proteintech
24377-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16872845
|
Proteintech
24377-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SETD9 is a protein-lysine N-methyltransferase. Monomethylates ′Lys-310′ of the RELA subunit of NF-kappa-B complex, leading to down-regulate NF-kappa-B transcription factor activity. Monomethylates ′Lys-8′ of H2AZ (H2AZK8me1). Required for the maintece of embryonic stem cell self-renewal.Specifications
SETD6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8TBK2, Q9CWY3 | |
66083, 79918 | |
SETD6 Fusion Protein Ag19485 | |
Primary | |
-20°C | |
SETD6 |
0.57 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
SET domain containing 6, SETD6 | |
Setd6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |