Läs mer
Invitrogen™ SETDB1 Monoclonal Antibody (5H6D4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515721
Beskrivning
MA5-15721 targets SETDB1 in indirect ELISA, WB applications and shows reactivity with Human, mouse, and Non-human primate samples. The MA5-15721 immunogen is purified recombinant fragment of human SETDB1 expressed in E. Coli. MA5-15721 detects SETDB1 which has a predicted molecular weight of approximately 180kDa.
This gene encodes a histone methyltransferase. The encoded enzyme catalyzes the reaction of S-adenosyl-L-methionine and histone L-lysine to produce S-adenosyl-L-homocysteine and histone N(6)-methyl-L-lysine. The encoded protein likely functions in transcriptional repression. Alternatively spliced transcript variants have been described.
Specifikationer
| SETDB1 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| O88974, Q15047 | |
| SETDB1 | |
| Purified recombinant fragment of human SETDB1 expressed in E. Coli. | |
| RUO | |
| 84505, 9869 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 5H6D4 | |
| Unconjugated | |
| SETDB1 | |
| AU022152; ERG-associated protein with a SET domain, ESET; ERG-associated protein with SET domain; ESET; H3-K9-HMTase 4; H3-K9-HMTase4; histone H3-K9 methyltransferase 4; Histone-lysine N-methyltransferase SETDB1; histone-lysine N-methyltransferase, H3lysine-9 specific 4; KG1T; KIAA0067; KMT1E; Lysine N-methyltransferase 1E; mKIAA0067; SET domain bifurcated 1; SET domain ERG-associated histone methyltransferase; SET domain, bifurcated 1; SETDB1; TDRD21; tudor domain containing 21 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG1 |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.