Läs mer
Setiptiline, TRC
Produktkod. 30409955
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30409955 5 mg
1 options
Produktkod. 30409955 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS2808005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Setiptiline
CAS 57262-94-9
Molekylformel C19H19N
Kvantitet 5 mg
Synonym 2,3,4,9-Tetrahydro-2-methyl-1H-dibenzo[3,4:6,7]cyclohepta[1,2-c]pyridine, Org 8282, MO-8282, Teciptilline
InChI formel InChI=1S/C19H19N/c1-20-11-10-18-16-8-4-2-6-14(16)12-15-7-3-5-9-17(15)19(18)13-20/h2-9H,10-13H2,1H3
LEDER CN1CCC2=C(C1)c3ccccc3Cc4ccccc24
Molekylvikt (g/mol) 261.36
Formel vikt 261.1517