Läs mer
Seviteronel, MedChemExpress
Produktkod. 16486958
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16486958 10 mg -
16453017 50 mg -
16447036 100 mg -
16439507 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16486958 Leverantör Medchem Express Leverantörsnummer HY15996
MedChemExpress Seviteronel (VT-464) is a potent CYP17 lyase inhibitor(h-Lyase IC50=69 nM) that demonstrated both exceptional in vitro lyase/hydroxylase selectivity (∼10-fold) and oral activity in a hamster model of androgen biosynthesis inhibition.
Seviteronel (VT-464) is a potent CYP17 lyase inhibitor(h-Lyase IC50=69 nM) that demonstrated both exceptional in vitro lyase/hydroxylase selectivity (∼10-fold) and oral activity in a hamster model of androgen biosynthesis inhibition.
Specifikationer
Kemiskt namn eller material Seviteronel
CAS 1610537-15-9
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C18H17F4N3O3
Kvantitet 10 mg
Synonym VT-464
Löslighetsinformation DMSO : ≥ 50 mg/mL (125.21 mM)
LEDER FC(F)OC1=C(OC(F)F)C=C(C=CC([C@](C2=CNN=N2)(C(C)C)O)=C3)C3=C1
Molekylvikt (g/mol) 399.34
Formel vikt 399.34
Procent renhet 95.65%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death