missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFPQ Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15585-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
SFPQ, also named PSF, encodes a nuclear factor implicated in the splicing and regulation of gene expression. SFPQ probably forms a heteromer with NONO and participates in DNA pairing and DNA break repair program. Very recently SFPQ was identified as a downstream target of tau, complete nuclear depletion and cytoplasmic accumulation of SFPQ were shown in the neurons and astrocytes of brains with Alzheimer′s disease (AD), more strikingly, reduced SFPQ levels may progress together with tau pathology, these observation strongly suggests the important role of SFPQ pathology in neurodegenerative diseases including AD. SFPQ protein may appears as 47 kDa, 52 kDa or 100 kDa molecules in various cell types.Specifications
SFPQ | |
Polyclonal | |
Unconjugated | |
SFPQ | |
100 kDa DNA pairing protein, hPOMp100, POMP100, PSF, PTB associated splicing factor, SFPQ | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
252855, 6421, 71514 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P23246, Q8VIJ6 | |
SFPQ | |
SFPQ Fusion Protein Ag7181 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |