Learn More
SFTPA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11850-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a lung surfactant protein that is a member of a subfamily of C-type lectins called collectins. The encoded protein binds specific carbohydrate moieties found on lipids and on the surface of microorganisms. This protein plays an essential role in surfactant homeostasis and in the defense against respiratory pathogens. Mutations in this gene are associated with idiopathic pulmonary fibrosis. Alternate splicing results in multiple transcript variants.Specifications
SFTPA1 | |
Polyclonal | |
Unconjugated | |
SFTPA1 | |
Alveolar proteinosis protein, COLEC4, Collectin 4, PSAP, PSP A, PSPA, SFTP1, SFTPA, SFTPA1, SFTPA1B, SP A, SP A1, Surfactant Protein A, surfactant protein A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
653509 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IWL2 | |
SFTPA1 | |
SFTPA1 Fusion Protein Ag2417 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |