missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTPA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SFTPA1 |
---|---|
Concentration | 0.53 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879193
|
Proteintech
11850-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16869193
|
Proteintech
11850-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a lung surfactant protein that is a member of a subfamily of C-type lectins called collectins. The encoded protein binds specific carbohydrate moieties found on lipids and on the surface of microorganisms. This protein plays an essential role in surfactant homeostasis and in the defense against respiratory pathogens. Mutations in this gene are associated with idiopathic pulmonary fibrosis. Alternate splicing results in multiple transcript variants.Specifications
SFTPA1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q8IWL2 | |
653509 | |
SFTPA1 Fusion Protein Ag2417 | |
Primary | |
-20°C | |
SFTPA1 |
0.53 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Alveolar proteinosis protein, COLEC4, Collectin 4, PSAP, PSP A, PSPA, SFTP1, SFTPA, SFTPA1, SFTPA1B, SP A, SP A1, Surfactant Protein A, surfactant protein A1 | |
SFTPA1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |