missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTPB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13664-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
ProSP-B is glycosylated in the Golgi apparatus and undergoes carboxy- and amino-terminal proteolysis by a cathepsin D-like protease. Pulmonary surfactant is a complex mixture of phospholipids and proteins that is secreted from type II cells in alveoli and reduces the surface tension at the alveolar air-liquid interface, providing alveolar stability necessary for normal ventilation. Four distinct proteins isolated from pulmonary surfactant are termed surfactant proteins A, B, C, and D. SP-A (28-36 kDa) and SP-D (43 kDa) are collagenous carbohydrate-binding proteins, whereas SP-B (8-9 kDa) and SP-C (4 kDa) are non-collagenous hydrophobic proteins. SP-B is expressed in pulmonary adenocarcinomas with acinar, papillary, bronchioloalveolar, and solid growth patterns. Squamous cell and large cell carcinomas of the lung and nonpulmonary adenocarcinomas do not express SP-B.Specifications
SFTPB | |
Polyclonal | |
Unconjugated | |
SFTPB | |
6 kDa protein, PSP B, SFTB3, SFTP3, SFTPB, SMDP1, SP B, surfactant protein B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6439 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P07988 | |
SFTPB | |
SFTPB Fusion Protein Ag4576 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |