missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFXN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SFXN3 |
---|---|
Concentration | 0.7 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16834214
|
Proteintech
15156-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16824214
|
Proteintech
15156-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Potential iron transporter.Specifications
SFXN3 | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q91V61, Q9BWM7, Q9JHY2 | |
65042, 81855, 94280 | |
SFXN3 Fusion Protein Ag7203 | |
Primary | |
-20°C | |
SFXN3 |
0.7 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
SFX3, SFXN3, sideroflexin 3 | |
Sfxn3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |