missing translation for 'onlineSavingsMsg'
Learn More

SFXN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15156-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824214

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potential iron transporter.
Specifications

Specifications

SFXN3
0.7 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q91V61, Q9BWM7, Q9JHY2
SFX3, SFXN3, sideroflexin 3
Rabbit
IgG
150 μL
-20°C
Primary
65042, 81855, 94280
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SFXN3
Liquid
Sfxn3
SFXN3 Fusion Protein Ag7203
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers