Läs mer
Tocris Bioscience™ SGC GAK 1
Beskrivning
SGC GAK 1 is a high affinity cyclin G associated kinase (GAK) inhibitor (Kd = 1.9 nM). Exhibits >30-fold selectivity for GAK over a panel of other kinases. Inhibits GAK in live cell assays (IC50 = 110 nM).
Specifikationer
Specifikationer
| Kemiskt namn eller material | 6-Bromo-N-(3,4,5-trimethoxyphenyl)-4-quinolinamine |
| CAS | 2226517-76-4 |
| Kvantitet | 25 mg |
| Molekylformel | C18H17BrN2O3 |
| Renhet | 99% |