Learn More
Abnova™ SGCA (Human) Recombinant Protein
Human SGCA full-length ORF ( NP_000014.1, 1 a.a. - 387 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00006442-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAETLFWTPLLVVLLAGLGDTEAQQTTLHPLVGRVFVHTLDHETFLSLPEHVAVPPAVHITYHAHLQGHPDLPRWLRYTQRSPHHPGFLYGSATPEDRGLQVIEVTAYNRDSFDTTRQRLVLEIGDPEGPLLPYQAEFLVRSHDAEEVLPSTPASRFLSALGGLWEPGELQLLNVTSALDRGGRVPLPIEGRKEGVYIKVGSASPFSTCLKMVASPDSHARCAQGQPPLLSCYDTLAPHFRVDWCNVTLVDKSVPEPADEVPTPGDGILEHDPFFCPPTEAPDRDFLVDALVTLLVPLLVALLLTLLLAYVMCCRREGRLKRDLATSDIQMVHHCTIHGNTEELRQMAASREVPRPLSTLPMFNVHTGERLPPRVDSAQVPLILDQH
Specifications
NP_000014.1 | |
sarcoglycan, alpha (50kDa dystrophin-associated glycoprotein) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SGCA | |
GST |
6442 | |
Wheat germ expression system | |
25 μg | |
50-DAG, A2, ADL, DAG2, DMDA2, LGMD2D, SCARMD1, adhalin | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |